Product Details

SNP ID
rs200231235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19876483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATCTTTCAATCATTGATATGTC[G/T]CTTGCTTCTTTCGCCACCCCAAAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4K2 PubMed Links

Gene Details

Gene
OR4K2
Gene Name
olfactory receptor family 4 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005501.1 216 Silent Mutation TCG,TCT S72S NP_001005501.1

View Full Product Details