Product Details

SNP ID
rs200660231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22946641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCCTCCCCAAGGACCTCATAGG[A/G]GTTCAGGACCTGTCTTGAGTTCTCC
Phenotype
MIM: 613431
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAUS4 PubMed Links

Gene Details

Gene
HAUS4
Gene Name
HAUS augmin like complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166269.1 Intron NP_001159741.1
NM_001166270.1 Intron NP_001159742.1
NM_017815.2 Intron NP_060285.2
Gene
LOC101926933
Gene Name
uncharacterized LOC101926933
There are no transcripts associated with this gene.

View Full Product Details