Product Details

SNP ID
rs201351353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58409104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGCTGAATATGATATTCCTGA[A/C]ATCTCATGGATATTCTTTGTGTCAT
Phenotype
MIM: 607384
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TIMM9 PubMed Links

Gene Details

Gene
TIMM9
Gene Name
translocase of inner mitochondrial membrane 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304485.1 726 Missense Mutation TGT,TTT C67F NP_001291414.1
NM_001304486.1 726 Missense Mutation TGT,TTT C67F NP_001291415.1
NM_001304487.1 726 Missense Mutation TGT,TTT C67F NP_001291416.1
NM_001304488.1 726 Intron NP_001291417.1
NM_001304489.1 726 Intron NP_001291418.1
NM_001304490.1 726 Intron NP_001291419.1
NM_001304491.1 726 Intron NP_001291420.1
NM_012460.3 726 Missense Mutation TGT,TTT C67F NP_036592.1
Gene
TOMM20L
Gene Name
translocase of outer mitochondrial membrane 20 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207377.2 726 Intron NP_997260.1
XM_011536742.2 726 Intron XP_011535044.1
XM_011536743.2 726 Intron XP_011535045.1
XM_011536744.2 726 Intron XP_011535046.1

View Full Product Details