Product Details

SNP ID
rs201820056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24367068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGGGAGAAAATGATAACCACCC[G/T]CCCATCTCTCCTACCCGCCAGCCTC
Phenotype
MIM: 602699
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NFATC4 PubMed Links

Gene Details

Gene
NFATC4
Gene Name
nuclear factor of activated T-cells 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136022.2 158 Missense Mutation CGC,CTC R5L NP_001129494.1
NM_001198965.1 158 Intron NP_001185894.1
NM_001198966.2 158 Intron NP_001185895.1
NM_001198967.2 158 Missense Mutation CGC,CTC R5L NP_001185896.1
NM_001288802.1 158 Intron NP_001275731.1
NM_001320043.1 158 Missense Mutation CGC,CTC R5L NP_001306972.1
NM_004554.4 158 Intron NP_004545.2
XM_011536797.2 158 Intron XP_011535099.1
XM_011536798.1 158 Intron XP_011535100.1
XM_011536799.2 158 Intron XP_011535101.1

View Full Product Details