Product Details

SNP ID
rs202187453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20556630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATTTACACGCTGGTATTTCAAAT[G/T]CTTCTGTTAAATTACAATACACTCC
Phenotype
MIM: 614014
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RNASE9 PubMed Links

Gene Details

Gene
RNASE9
Gene Name
ribonuclease A family member 9 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001673.3 600 Missense Mutation GAA,GCA E147A NP_001001673.2
NM_001110356.1 600 Missense Mutation GAA,GCA E147A NP_001103826.1
NM_001110357.1 600 Missense Mutation GAA,GCA E147A NP_001103827.1
NM_001110358.1 600 Missense Mutation GAA,GCA E152A NP_001103828.1
NM_001110359.1 600 Missense Mutation GAA,GCA E152A NP_001103829.1
NM_001110360.1 600 Missense Mutation GAA,GCA E152A NP_001103830.1
NM_001110361.1 600 Missense Mutation GAA,GCA E152A NP_001103831.1
NM_001289110.1 600 Missense Mutation GAA,GCA E147A NP_001276039.1

View Full Product Details