Product Details

SNP ID
rs201283346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:56095005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTGAAGGGTCATTCCCAAACAG[A/T]TGCCGTAAGAATGAGAATTGTTCAT
Phenotype
MIM: 612660
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RFX7 PubMed Links

Gene Details

Gene
RFX7
Gene Name
regulatory factor X7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022841.5 3952 Missense Mutation AAC,ATC N908I NP_073752.5
XM_005254603.3 3952 Missense Mutation AAC,ATC N908I XP_005254660.2
XM_011521925.2 3952 Missense Mutation AAC,ATC N811I XP_011520227.1
XM_017022506.1 3952 Missense Mutation AAC,ATC N908I XP_016877995.1
XM_017022507.1 3952 Missense Mutation AAC,ATC N811I XP_016877996.1
XM_017022508.1 3952 Missense Mutation AAC,ATC N811I XP_016877997.1

View Full Product Details