Product Details

SNP ID
rs200536870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45116149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCACTTCAGTGCAGAATGGTT[C/T]GTGGGTACAGTGAACACCAACACAT
Phenotype
MIM: 606759 MIM: 612771 MIM: 612772
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUOX2 PubMed Links

Gene Details

Gene
DUOX2
Gene Name
dual oxidase 2
There are no transcripts associated with this gene.

Gene
DUOXA1
Gene Name
dual oxidase maturation factor 1
There are no transcripts associated with this gene.

Gene
DUOXA2
Gene Name
dual oxidase maturation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207581.3 516 Silent Mutation TTC,TTT F77F NP_997464.2
XM_017022180.1 516 Missense Mutation TTC,TTT F77F XP_016877669.1

View Full Product Details