Product Details
- SNP ID
-
rs200736815
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:90969493 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAACAGAATTAATGCTGAAGTTG[C/T]GCTGGAGGGGGGCCGAGCCAGGGTA
- Phenotype
-
MIM: 603484
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRC1
PubMed Links
Gene Details
- Gene
- PRC1
- Gene Name
- protein regulator of cytokinesis 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001267580.1 |
1524 |
Intron |
|
|
NP_001254509.1 |
NM_003981.3 |
1524 |
Missense Mutation |
CAC,CGC |
H568R |
NP_003972.1 |
NM_199413.2 |
1524 |
Missense Mutation |
CAC,CGC |
H568R |
NP_955445.1 |
XM_005254987.2 |
1524 |
Intron |
|
|
XP_005255044.1 |
XM_006720759.1 |
1524 |
Intron |
|
|
XP_006720822.1 |
XM_006720760.1 |
1524 |
Intron |
|
|
XP_006720823.1 |
XM_011522187.1 |
1524 |
Missense Mutation |
CAC,CGC |
H568R |
XP_011520489.1 |
XM_011522188.2 |
1524 |
Missense Mutation |
CAC,CGC |
H568R |
XP_011520490.1 |
XM_011522189.1 |
1524 |
Missense Mutation |
CAC,CGC |
H531R |
XP_011520491.1 |
XM_011522190.2 |
1524 |
Missense Mutation |
CAC,CGC |
H511R |
XP_011520492.1 |
XM_011522191.2 |
1524 |
Intron |
|
|
XP_011520493.1 |
XM_011522192.1 |
1524 |
Missense Mutation |
CAC,CGC |
H461R |
XP_011520494.1 |
XM_017022712.1 |
1524 |
Missense Mutation |
CAC,CGC |
H531R |
XP_016878201.1 |
XM_017022713.1 |
1524 |
Missense Mutation |
CAC,CGC |
H531R |
XP_016878202.1 |
XM_017022714.1 |
1524 |
Missense Mutation |
CAC,CGC |
H516R |
XP_016878203.1 |
XM_017022715.1 |
1524 |
Missense Mutation |
CAC,CGC |
H516R |
XP_016878204.1 |
XM_017022716.1 |
1524 |
Missense Mutation |
CAC,CGC |
H511R |
XP_016878205.1 |
XM_017022717.1 |
1524 |
Intron |
|
|
XP_016878206.1 |
- Gene
- PRC1-AS1
- Gene Name
- PRC1 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- RCCD1
- Gene Name
- RCC1 domain containing 1
There are no transcripts associated with this gene.
View Full Product Details