Product Details

SNP ID
rs199806322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACTCACAGCGGACAGGATAGAC[C/G]TAGAAGAGGACCACCAGGCCAGCCA
Phenotype
MIM: 606007 MIM: 614403
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POLR3K PubMed Links

Gene Details

Gene
POLR3K
Gene Name
RNA polymerase III subunit K
There are no transcripts associated with this gene.

Gene
RHBDF1
Gene Name
rhomboid 5 homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022450.3 2679 Silent Mutation TAC,TAG Y823* NP_071895.3
XM_005255494.1 2679 Nonsense Mutation TAC,TAG Y823* XP_005255551.1
XM_005255498.2 2679 Silent Mutation TAC,TAG Y507* XP_005255555.1
XM_006720921.1 2679 Silent Mutation TAC,TAG Y846* XP_006720984.1
XM_017023556.1 2679 Silent Mutation TAC,TAG Y846* XP_016879045.1
XM_017023557.1 2679 Silent Mutation TAC,TAG Y792* XP_016879046.1
XM_017023558.1 2679 Intron XP_016879047.1
Gene
SNRNP25
Gene Name
small nuclear ribonucleoprotein U11/U12 subunit 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024571.3 2679 Intron NP_078847.1

View Full Product Details