Product Details

SNP ID
rs199977814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30445402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCCCAGGGCTGGAAAGGTGGGG[C/T]TGGGGCCCGCTCCTGCCGGCAGGCC
Phenotype
MIM: 606218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEPHS2 PubMed Links

Gene Details

Gene
SEPHS2
Gene Name
selenophosphate synthetase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012248.3 574 Missense Mutation AAC,AGC N109S NP_036380.2

View Full Product Details