Product Details

SNP ID
rs200835240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67163247 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGTGGGTCCCGGCTCAGTGCT[C/G]GAGCTACTGGAGGCCTGCCCGCGCC
Phenotype
MIM: 609077 MIM: 602438 MIM: 605235 MIM: 603500
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXL8 PubMed Links

Gene Details

Gene
FBXL8
Gene Name
F-box and leucine rich repeat protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018378.2 672 Silent Mutation CTC,CTG L184L NP_060848.2
Gene
HSF4
Gene Name
heat shock transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040667.2 672 Intron NP_001035757.1
NM_001538.3 672 Intron NP_001529.2
Gene
NOL3
Gene Name
nucleolar protein 3
There are no transcripts associated with this gene.

Gene
TRADD
Gene Name
TNFRSF1A associated via death domain
There are no transcripts associated with this gene.

View Full Product Details