Product Details

SNP ID
rs201420090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57447520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTTCCAAAATGGCGGCGGCGGC[A/G]GTATCTGGTGCGCTTGGCCGGGCGG
Phenotype
MIM: 608943 MIM: 612837
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIAPIN1 PubMed Links

Gene Details

Gene
CIAPIN1
Gene Name
cytokine induced apoptosis inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308347.1 96 Intron NP_001295276.1
NM_001308358.1 96 Intron NP_001295287.1
NM_020313.3 96 Intron NP_064709.2
XM_005256063.4 96 Intron XP_005256120.1
Gene
COQ9
Gene Name
coenzyme Q9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020312.3 96 Silent Mutation GCA,GCG A5A NP_064708.1

View Full Product Details