Product Details

SNP ID
rs200375017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3243364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGGAGGCTCCTTTATTAGCAGG[C/T]GGGTCGGGGGAACGCTGGACGCCTG
Phenotype
MIM: 608107 MIM: 603231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEFV PubMed Links

Gene Details

Gene
MEFV
Gene Name
Mediterranean fever
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000243.2 2301 Missense Mutation NP_000234.1
NM_001198536.1 2301 UTR 3 NP_001185465.1
XM_017023236.1 2301 Missense Mutation XP_016878725.1
Gene
ZNF200
Gene Name
zinc finger protein 200
There are no transcripts associated with this gene.

View Full Product Details