Product Details

SNP ID
rs199924156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3090473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCACTGCTGCCAAGAAGGGACC[C/T]ATCTTCACCAGGGCCGCTAGCCGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN10 PubMed Links

Gene Details

Gene
ZSCAN10
Gene Name
zinc finger and SCAN domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282415.1 888 UTR 5 NP_001269344.1
NM_001282416.1 888 Missense Mutation AGG,GGG R184G NP_001269345.1
NM_032805.2 888 Missense Mutation AGG,GGG R321G NP_116194.2
XM_017023791.1 888 Missense Mutation AGG,GGG R321G XP_016879280.1
XM_017023792.1 888 Missense Mutation AGG,GGG R321G XP_016879281.1

View Full Product Details