Product Details

SNP ID
rs201140031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30086387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCGGCAGCTCCAGAAATGCAGC[C/T]GAGTAGGGGGCTGAGCGCCCGGAGT
Phenotype
MIM: 602035 MIM: 602427 MIM: 609724
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP4C PubMed Links

Gene Details

Gene
PPP4C
Gene Name
protein phosphatase 4 catalytic subunit
There are no transcripts associated with this gene.

Gene
TBX6
Gene Name
T-box 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004608.3 1234 Silent Mutation TCA,TCG S383S NP_004599.2
XM_005255523.2 1234 Silent Mutation TCA,TCG S383S XP_005255580.1
XM_011545926.2 1234 Silent Mutation TCA,TCG S383S XP_011544228.1
XM_017023614.1 1234 Silent Mutation TCA,TCG S383S XP_016879103.1
Gene
YPEL3
Gene Name
yippee like 3
There are no transcripts associated with this gene.

View Full Product Details