Product Details

SNP ID
rs201613304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67291316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCCTGTCACACATACCCACAGC[C/T]GATTGGCCAGGGAGACCACCTTGGC
Phenotype
MIM: 609526
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD19 PubMed Links

Gene Details

Gene
KCTD19
Gene Name
potassium channel tetramerization domain containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100915.1 2609 Missense Mutation CAG,CGG Q853R NP_001094385.1
Gene
PLEKHG4
Gene Name
pleckstrin homology and RhoGEF domain containing G4
There are no transcripts associated with this gene.

View Full Product Details