Product Details

SNP ID
rs202134725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30445311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGATCTTCTACCAAGGGGTAAAA[G/T]AAGTCCGTGGTCTGCACCAGTGACA
Phenotype
MIM: 606218
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEPHS2 PubMed Links

Gene Details

Gene
SEPHS2
Gene Name
selenophosphate synthetase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012248.3 665 Silent Mutation NP_036380.2

View Full Product Details