Product Details

SNP ID
rs201593079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30953600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCAATCTCCCACGGTCCTCTG[C/T]GTCTGCAGCACCGTCCCAAGCTGAG
Phenotype
MIM: 609085 MIM: 610930 MIM: 611052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL19 PubMed Links

Gene Details

Gene
FBXL19
Gene Name
F-box and leucine rich repeat protein 19
There are no transcripts associated with this gene.

Gene
ORAI3
Gene Name
ORAI calcium release-activated calcium modulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152288.2 850 Missense Mutation GCG,GTG A215V NP_689501.1
Gene
SETD1A
Gene Name
SET domain containing 1A
There are no transcripts associated with this gene.

View Full Product Details