Product Details

SNP ID
rs200239930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4780817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTCCGGGGATTGCAGCCGTTC[A/G]GGCAGGGGGCAAGGGCTGCTGTGGT
Phenotype
MIM: 604657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLTPD2 PubMed Links

Gene Details

Gene
GLTPD2
Gene Name
glycolipid transfer protein domain containing 2
There are no transcripts associated with this gene.

Gene
TM4SF5
Gene Name
transmembrane 4 L six family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003963.2 237 Missense Mutation CAG,CGG Q69R NP_003954.2
Gene
VMO1
Gene Name
vitelline membrane outer layer 1 homolog
There are no transcripts associated with this gene.

View Full Product Details