Product Details

SNP ID
rs200049183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75899222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGGCACTGGAAGAAGGACAAGCC[A/G]GCTGGAGTCATGGTTTCTTGGTGTT
Phenotype
MIM: 611844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL38 PubMed Links

Gene Details

Gene
MRPL38
Gene Name
mitochondrial ribosomal protein L38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032478.3 1167 Silent Mutation GCC,GCT A314A NP_115867.2
Gene
TRIM65
Gene Name
tripartite motif containing 65
There are no transcripts associated with this gene.

View Full Product Details