Product Details

SNP ID
rs200479403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39742370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACCTTTGCCCAGCCCCTGGGAC[A/C]CCTCCTGGGACTCCCCGGCCCCCTG
Phenotype
MIM: 601522
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GRB7 PubMed Links

Gene Details

Gene
GRB7
Gene Name
growth factor receptor bound protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030002.2 334 Silent Mutation ACA,ACC T23T NP_001025173.1
NM_001242442.1 334 Silent Mutation ACA,ACC T46T NP_001229371.1
NM_001242443.1 334 Silent Mutation ACA,ACC T23T NP_001229372.1
NM_005310.3 334 Silent Mutation ACA,ACC T23T NP_005301.2
XM_017024536.1 334 UTR 5 XP_016880025.1
XM_017024537.1 334 Silent Mutation ACA,ACC T23T XP_016880026.1
XM_017024538.1 334 UTR 5 XP_016880027.1

View Full Product Details