Product Details

SNP ID
rs202125336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29043294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGATCCAGGGCTGCTTCACTGC[A/G]TACCACCTGGCCAAACACAGGAAGA
Phenotype
MIM: 616713
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIPOX PubMed Links

Gene Details

Gene
PIPOX
Gene Name
pipecolic acid and sarcosine oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016518.2 395 Silent Mutation GCA,GCG A23A NP_057602.2
XM_017024722.1 395 Intron XP_016880211.1

View Full Product Details