Product Details

SNP ID
rs201545163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3897851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTTCTCCTGCAGGCCCAGGGTG[A/G]AGCTGGTAGCTGCGAGGTCACGCTC
Phenotype
MIM: 611411 MIM: 600845
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMKK1 PubMed Links

Gene Details

Gene
CAMKK1
Gene Name
calcium/calmodulin dependent protein kinase kinase 1
There are no transcripts associated with this gene.

Gene
P2RX1
Gene Name
purinergic receptor P2X 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002558.3 1604 Missense Mutation TCC,TTC S388F NP_002549.1
XM_006721529.2 1604 Missense Mutation TCC,TTC S371F XP_006721592.1
XM_011523896.2 1604 Missense Mutation TCC,TTC S420F XP_011522198.1
XM_011523897.2 1604 Missense Mutation TCC,TTC S408F XP_011522199.1
XM_011523898.2 1604 Missense Mutation TCC,TTC S403F XP_011522200.1
XM_011523899.2 1604 Missense Mutation TCC,TTC S400F XP_011522201.1
XM_011523900.2 1604 Intron XP_011522202.1

View Full Product Details