Product Details

SNP ID
rs202104033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35920198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCTTCACATACTTGAATTAAAC[C/T]GTGTAGTTCTTCTTCGCATCTGACA
Phenotype
MIM: 612896
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC37A8P PubMed Links

Gene Details

Gene
LRRC37A8P
Gene Name
leucine rich repeat containing 37 member A8, pseudogene
There are no transcripts associated with this gene.

Gene
RDM1
Gene Name
RAD52 motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034836.1 692 Intron NP_001030008.1
NM_001163120.1 692 Intron NP_001156592.1
NM_001163121.1 692 Missense Mutation AGT,GGT S215G NP_001156593.1
NM_001163122.1 692 Missense Mutation AGT,GGT S192G NP_001156594.1
NM_001163124.1 692 Intron NP_001156596.1
NM_001163125.1 692 Intron NP_001156597.1
NM_001163130.1 692 Missense Mutation AGT,GGT S225G NP_001156602.1
NM_145654.3 692 Missense Mutation AGT,GGT S248G NP_663629.1
XM_011524509.2 692 Intron XP_011522811.1
XM_011524510.2 692 Intron XP_011522812.1

View Full Product Details