Product Details

SNP ID
rs202173320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44006162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGACTCCCTGGAGGTGACCGCGTC[A/G]CTGGCCAAGGCGCTGCGGCCCACCA
Phenotype
MIM: 608300 MIM: 600781
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAGS PubMed Links

Gene Details

Gene
NAGS
Gene Name
N-acetylglutamate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153006.2 730 Silent Mutation TCA,TCG S280S NP_694551.1
XM_011524438.1 730 Silent Mutation TCA,TCG S280S XP_011522740.1
XM_011524439.2 730 Silent Mutation TCA,TCG S114S XP_011522741.1
Gene
PYY
Gene Name
peptide YY
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004160.5 730 Intron NP_004151.3
XM_011525035.1 730 Intron XP_011523337.1
Gene
TMEM101
Gene Name
transmembrane protein 101
There are no transcripts associated with this gene.

View Full Product Details