Product Details
- SNP ID
-
rs200833034
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:8121844 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGCGCTGGAGGCCTCGGATCTAC[C/T]GGCTTGGGCCGGGGCGGTTTGGGGC
- Phenotype
-
MIM: 607206
MIM: 608059
MIM: 610407
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALOXE3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56066045] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALOXE3
- Gene Name
- arachidonate lipoxygenase 3
There are no transcripts associated with this gene.
- Gene
- HES7
- Gene Name
- hes family bHLH transcription factor 7
- Gene
- TRG-GCC2-6
- Gene Name
- transfer RNA-Gly (GCC) 2-6
There are no transcripts associated with this gene.
View Full Product Details