Product Details
- SNP ID
-
rs201016237
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40878262 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGCGTTCCATGTTACTCCGAATC[C/T]GCATCAGTTGGGCCTCCAGAGAGCT
- Phenotype
-
MIM: 608218
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT20
PubMed Links
Gene Details
- Gene
- KRT20
- Gene Name
- keratin 20
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019010.2 |
1080 |
Missense Mutation |
CAG,CGG |
Q341R |
NP_061883.1 |
View Full Product Details