Product Details

SNP ID
rs201975637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64858856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGCCAAGATGAGTTTTTTGGTAT[A/G]GCCATATCCTGGAAGTTCTTTTGTG
Phenotype
MIM: 616557
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC37A3 PubMed Links

Gene Details

Gene
LRRC37A3
Gene Name
leucine rich repeat containing 37 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303255.1 5398 Missense Mutation NP_001290184.1
NM_199340.3 5398 Missense Mutation NP_955372.2
XM_011524776.1 5398 Intron XP_011523078.1
XM_011524777.1 5398 Intron XP_011523079.1
XM_011524778.2 5398 Intron XP_011523080.2
XM_017024593.1 5398 Intron XP_016880082.1
XM_017024594.1 5398 Intron XP_016880083.1
XM_017024595.1 5398 Intron XP_016880084.1
XM_017024596.1 5398 Intron XP_016880085.1
XM_017024597.1 5398 Intron XP_016880086.1
XM_017024598.1 5398 Intron XP_016880087.1
XM_017024599.1 5398 Intron XP_016880088.1
XM_017024600.1 5398 Intron XP_016880089.1
XM_017024601.1 5398 Missense Mutation CAT,TAT H842Y XP_016880090.1
XM_017024602.1 5398 Missense Mutation CAT,TAT H767Y XP_016880091.1
XM_017024603.1 5398 Intron XP_016880092.1
XM_017024604.1 5398 Intron XP_016880093.1
XM_017024605.1 5398 Intron XP_016880094.1
XM_017024606.1 5398 Intron XP_016880095.1
XM_017024607.1 5398 Intron XP_016880096.1
XM_017024608.1 5398 Intron XP_016880097.1

View Full Product Details