Product Details

SNP ID
rs200494000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:36114342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCATTTATTCTTGCACCCTCTG[A/G]GTACATATTCATTGTAGACTTCCTG
Phenotype
MIM: 608731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC39A6 PubMed Links

Gene Details

Gene
SLC39A6
Gene Name
solute carrier family 39 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099406.1 2321 Missense Mutation CCC,CTC P258L NP_001092876.1
NM_012319.3 2321 Missense Mutation CCC,CTC P533L NP_036451.3
XM_011525900.1 2321 Missense Mutation CCC,CTC P533L XP_011524202.1
XM_011525901.1 2321 Missense Mutation CCC,CTC P533L XP_011524203.1

View Full Product Details