Product Details

SNP ID
rs200009740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5891530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCGCGTTTGCGCAGATGAAGAG[A/G]AAGATGCCGATGCCCATGATGAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM200C PubMed Links

Gene Details

Gene
TMEM200C
Gene Name
transmembrane protein 200C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080209.1 575 Silent Mutation TTC,TTT F178F NP_001073678.1
XM_005258130.4 575 Silent Mutation TTC,TTT F178F XP_005258187.1
XM_011525729.2 575 Silent Mutation TTC,TTT F178F XP_011524031.1

View Full Product Details