Product Details

SNP ID
rs200719522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:58482033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGGAACAGTTGCCTTTAAATCC[C/T]TTGTACCTGTGAGTTTGGGTGGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALPK2 PubMed Links

Gene Details

Gene
ALPK2
Gene Name
alpha kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052947.3 5635 Silent Mutation NP_443179.3
XM_011525801.2 5635 Missense Mutation XP_011524103.1
XM_011525802.2 5635 Missense Mutation XP_011524104.1
XM_017025535.1 5635 Intron XP_016881024.1
XM_017025536.1 5635 Intron XP_016881025.1
XM_017025537.1 5635 Intron XP_016881026.1
XM_017025538.1 5635 Missense Mutation XP_016881027.1
XM_017025539.1 5635 Missense Mutation XP_016881028.1

View Full Product Details