Product Details

SNP ID
rs201451060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:7231165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGGCCAGGCAGTCAAGGGCCAG[C/T]TCCAAGGATAAGGGCCCCAAGAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC30 PubMed Links

Gene Details

Gene
LRRC30
Gene Name
leucine rich repeat containing 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105581.1 27 Silent Mutation AGC,AGT S9S NP_001099051.1

View Full Product Details