Product Details

SNP ID
rs200340957
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38405576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCAGTACCGCTGCCGCCGCTGC[C/T]TCCTCCTCAAGCGCCTTGACCTCAC
Phenotype
MIM: 607320 MIM: 609293
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM98C PubMed Links
Additional Information
For this assay, SNP(s) [rs3745962] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FAM98C
Gene Name
family with sequence similarity 98 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174905.3 710 Missense Mutation CTC,TTC L231F NP_777565.3
XM_017026354.1 710 UTR 3 XP_016881843.1
Gene
RASGRP4
Gene Name
RAS guanyl releasing protein 4
There are no transcripts associated with this gene.

Gene
SPRED3
Gene Name
sprouty related EVH1 domain containing 3
There are no transcripts associated with this gene.

View Full Product Details