Product Details

SNP ID
rs200141569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55378088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGAGGGACCAAGAAGACGCCGT[C/G]CACGGGCAGCGTGCCAGTCGCCCTG
Phenotype
MIM: 147681 MIM: 603638
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL11 PubMed Links

Gene Details

Gene
IL11
Gene Name
interleukin 11
There are no transcripts associated with this gene.

Gene
RPL28
Gene Name
ribosomal protein L28
There are no transcripts associated with this gene.

Gene
TMEM190
Gene Name
transmembrane protein 190
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139172.1 437 Missense Mutation TCC,TGC S140C NP_631911.1
XM_017026331.1 437 Missense Mutation TCC,TGC S98C XP_016881820.1
Gene
TMEM238
Gene Name
transmembrane protein 238
There are no transcripts associated with this gene.

View Full Product Details