Product Details

SNP ID
rs199758385
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38405080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCAGACCTGCACCAGGGACCCC[C/T]GCCAGCCAGCTGCTGCAGGAGTTGC
Phenotype
MIM: 607320 MIM: 609293
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM98C PubMed Links

Gene Details

Gene
FAM98C
Gene Name
family with sequence similarity 98 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174905.3 541 Silent Mutation CCC,CCT P174P NP_777565.3
XM_017026354.1 541 Silent Mutation CCC,CCT P174P XP_016881843.1
Gene
RASGRP4
Gene Name
RAS guanyl releasing protein 4
There are no transcripts associated with this gene.

Gene
SPRED3
Gene Name
sprouty related EVH1 domain containing 3
There are no transcripts associated with this gene.

View Full Product Details