Product Details

SNP ID
rs200322282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3589517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGACCAGCGGGAGGGAGACCCTG[C/T]GGCTTCGTTCTGGGGGGGCTGCCAC
Phenotype
MIM: 608792 MIM: 188070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIPC3 PubMed Links

Gene Details

Gene
GIPC3
Gene Name
GIPC PDZ domain containing family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133261.2 734 Missense Mutation CGG,TGG R223W NP_573568.1
XM_005259492.3 734 Missense Mutation CGG,TGG R223W XP_005259549.1
Gene
TBXA2R
Gene Name
thromboxane A2 receptor
There are no transcripts associated with this gene.

View Full Product Details