Product Details

SNP ID
rs200049098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48625977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGGCCATGGCCCCGCCCCCACC[A/G]CCACTGGCTGCCAGCACCCCGCTCC
Phenotype
MIM: 124097 MIM: 604179 MIM: 607092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DBP PubMed Links

Gene Details

Gene
DBP
Gene Name
D-box binding PAR bZIP transcription factor
There are no transcripts associated with this gene.

Gene
RPL18
Gene Name
ribosomal protein L18
There are no transcripts associated with this gene.

Gene
SPHK2
Gene Name
sphingosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204158.2 182 Intron NP_001191087.1
NM_001204159.2 182 Silent Mutation CCA,CCG P42P NP_001191088.1
NM_001204160.2 182 Silent Mutation CCA,CCG P6P NP_001191089.1
NM_001243876.1 182 Intron NP_001230805.1
NM_020126.4 182 Silent Mutation CCA,CCG P42P NP_064511.2
XM_006723292.1 182 Silent Mutation CCA,CCG P6P XP_006723355.1
XM_011527133.1 182 Silent Mutation CCA,CCG P42P XP_011525435.1
XM_011527134.1 182 Silent Mutation CCA,CCG P6P XP_011525436.1
XM_017027008.1 182 Silent Mutation CCA,CCG P104P XP_016882497.1
XM_017027009.1 182 Intron XP_016882498.1
XM_017027010.1 182 Silent Mutation CCA,CCG P42P XP_016882499.1

View Full Product Details