Product Details

SNP ID
rs199527137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7847741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTTCCTGCCCAGGCTCCTGGA[A/G]GCCGACTCCAAGTCCATGCGCTCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EVI5L PubMed Links

Gene Details

Gene
EVI5L
Gene Name
ecotropic viral integration site 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159944.2 287 Silent Mutation GAA,GAG E49E NP_001153416.1
NM_145245.4 287 Silent Mutation GAA,GAG E49E NP_660288.1
XM_005272458.4 287 Silent Mutation GAA,GAG E49E XP_005272515.1
XM_017026239.1 287 Silent Mutation GAA,GAG E49E XP_016881728.1

View Full Product Details