Product Details

SNP ID
rs200472617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45012268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGAGGCCAGCAAGACGCTGCCC[A/G]CCATCGAGGTGGGCCCGGCGAGCGG
Phenotype
MIM: 604758
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RELB PubMed Links

Gene Details

Gene
RELB
Gene Name
RELB proto-oncogene, NF-kB subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006509.3 627 Missense Mutation ACC,GCC T166A NP_006500.2
XM_005259127.3 627 Missense Mutation ACC,GCC T163A XP_005259184.1
XM_005259128.2 627 Missense Mutation ACC,GCC T166A XP_005259185.1

View Full Product Details