Product Details

SNP ID
rs200650622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2876112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAGATAATGAGGCTCAGCTTAA[A/T]GCCAGTGGGTCTATTTCTCAGCAGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF556 PubMed Links

Gene Details

Gene
ZNF556
Gene Name
zinc finger protein 556
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300843.1 237 Missense Mutation AAA,AAT K50N NP_001287772.1
NM_024967.2 237 Missense Mutation AAA,AAT K50N NP_079243.1

View Full Product Details