Product Details

SNP ID
rs56184317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54813863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGACTCTGATGAACAAGACCCT[C/G]AGGAGGTGACATACGCACAGTTGGA
Phenotype
MIM: 604945 MIM: 604946
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIR2DL4 PubMed Links

Gene Details

Gene
KIR2DL4
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080770.1 925 Missense Mutation CAG,GAG Q261E NP_001074239.1
NM_001080772.1 925 UTR 3 NP_001074241.1
NM_002255.5 925 Missense Mutation CAG,GAG Q296E NP_002246.5
Gene
KIR3DL1
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 1
There are no transcripts associated with this gene.

Gene
LOC102724300
Gene Name
uncharacterized LOC102724300
There are no transcripts associated with this gene.

View Full Product Details