Product Details

SNP ID
rs202128704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49593342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCCCAGCTTGGTTTATGGCAG[C/G]TCCAGGACCTCGCACCCCGAGACGG
Phenotype
MIM: 616633 MIM: 604429
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRR12 PubMed Links

Gene Details

Gene
PRR12
Gene Name
proline rich 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017027031.1 580 Missense Mutation AGC,AGG S34R XP_016882520.1
Gene
PRRG2
Gene Name
proline rich and Gla domain 2
There are no transcripts associated with this gene.

View Full Product Details