Product Details
- SNP ID
-
rs201812504
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51993292 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTCCACATTCATTGCATATATAA[A/G]GTTTCTCCCCAGTATGAGTTCGCTG
- Phenotype
-
MIM: 605422
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF350
PubMed Links
Gene Details
- Gene
- ZNF350
- Gene Name
- zinc finger protein 350
There are no transcripts associated with this gene.
- Gene
- ZNF615
- Gene Name
- zinc finger protein 615
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199324.1 |
2134 |
Missense Mutation |
CCT,CTT |
P606L |
NP_001186253.1 |
NM_001321317.1 |
2134 |
Missense Mutation |
CCT,CTT |
P600L |
NP_001308246.1 |
NM_001321318.1 |
2134 |
Missense Mutation |
CCT,CTT |
P602L |
NP_001308247.1 |
NM_001321319.1 |
2134 |
Missense Mutation |
CCT,CTT |
P606L |
NP_001308248.1 |
NM_001321320.1 |
2134 |
Missense Mutation |
CCT,CTT |
P595L |
NP_001308249.1 |
NM_001321321.1 |
2134 |
Missense Mutation |
CCT,CTT |
P606L |
NP_001308250.1 |
NM_001321322.1 |
2134 |
Missense Mutation |
CCT,CTT |
P602L |
NP_001308251.1 |
NM_001321323.1 |
2134 |
Missense Mutation |
CCT,CTT |
P611L |
NP_001308252.1 |
NM_198480.3 |
2134 |
Missense Mutation |
CCT,CTT |
P595L |
NP_940882.3 |
XM_011526824.1 |
2134 |
Missense Mutation |
CCT,CTT |
P606L |
XP_011525126.1 |
XM_011526825.1 |
2134 |
Missense Mutation |
CCT,CTT |
P606L |
XP_011525127.1 |
XM_011526826.1 |
2134 |
Missense Mutation |
CCT,CTT |
P606L |
XP_011525128.1 |
XM_011526829.1 |
2134 |
Missense Mutation |
CCT,CTT |
P487L |
XP_011525131.1 |
XM_011526830.1 |
2134 |
Missense Mutation |
CCT,CTT |
P487L |
XP_011525132.1 |
XM_017026648.1 |
2134 |
Missense Mutation |
CCT,CTT |
P595L |
XP_016882137.1 |
XM_017026649.1 |
2134 |
Missense Mutation |
CCT,CTT |
P595L |
XP_016882138.1 |
XM_017026650.1 |
2134 |
Missense Mutation |
CCT,CTT |
P487L |
XP_016882139.1 |
XM_017026651.1 |
2134 |
Missense Mutation |
CCT,CTT |
P487L |
XP_016882140.1 |
View Full Product Details