Product Details

SNP ID
rs201654008
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:40541696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGGTGGAGGGAGCGAATCCAC[A/G]CGAGAGCTCTCCAGGTGAGTGAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF684 PubMed Links

Gene Details

Gene
ZNF684
Gene Name
zinc finger protein 684
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152373.3 808 Missense Mutation CAC,CGC H75R NP_689586.3
XM_011540671.2 808 Missense Mutation CAC,CGC H75R XP_011538973.1
XM_011540672.2 808 Missense Mutation CAC,CGC H75R XP_011538974.1
XM_011540673.2 808 Missense Mutation CAC,CGC H75R XP_011538975.1
XM_017000290.1 808 Missense Mutation CAC,CGC H75R XP_016855779.1
XM_017000291.1 808 Missense Mutation CAC,CGC H39R XP_016855780.1
XM_017000292.1 808 Missense Mutation CAC,CGC H8R XP_016855781.1

View Full Product Details