Product Details

SNP ID
rs201192220
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:159952337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTGTTGGCTCTGCAGGTGTAGG[A/C]GAGGGCACTGTCCCCCGGCCTCCAG
Phenotype
MIM: 609738 MIM: 608589
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGSF9 PubMed Links

Gene Details

Gene
IGSF9
Gene Name
immunoglobulin superfamily member 9
There are no transcripts associated with this gene.

Gene
LINC01133
Gene Name
long intergenic non-protein coding RNA 1133
There are no transcripts associated with this gene.

Gene
SLAMF9
Gene Name
SLAM family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146172.1 706 Intron NP_001139644.1
NM_001146173.1 706 Intron NP_001139645.1
NM_033438.3 706 Missense Mutation GCC,TCC A197S NP_254273.2
XM_017002756.1 706 Missense Mutation GCC,TCC A227S XP_016858245.1

View Full Product Details