Product Details

SNP ID
rs201614228
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:20143474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCACAGCTCACGGCAGCCTGCT[C/T]AACCTGAAGGCCATGGTGGAGGCCG
Phenotype
MIM: 616793
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G2F PubMed Links

Gene Details

Gene
PLA2G2F
Gene Name
phospholipase A2 group IIF
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022819.3 289 Silent Mutation CTC,CTT L66L NP_073730.3
XM_011541955.1 289 Silent Mutation CTC,CTT L66L XP_011540257.1
XM_011541956.1 289 Silent Mutation CTC,CTT L66L XP_011540258.1

View Full Product Details