Product Details
- SNP ID
-
rs200994363
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:243270244 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGACAGCCTGGCCCGAATTACA[A/G]CAGAGCCATGCTGGTGAGTGTGAAT
- Phenotype
-
MIM: 613524
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SDCCAG8
PubMed Links
Gene Details
- Gene
- SDCCAG8
- Gene Name
- serologically defined colon cancer antigen 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006642.3 |
344 |
Silent Mutation |
CAA,CAG |
Q69Q |
NP_006633.1 |
XM_005273013.4 |
344 |
Silent Mutation |
CAA,CAG |
Q69Q |
XP_005273070.1 |
XM_005273018.1 |
344 |
UTR 5 |
|
|
XP_005273075.1 |
XM_005273021.4 |
344 |
UTR 5 |
|
|
XP_005273078.1 |
XM_005273022.3 |
344 |
UTR 5 |
|
|
XP_005273079.1 |
XM_005273023.4 |
344 |
Silent Mutation |
CAA,CAG |
Q69Q |
XP_005273080.1 |
XM_006711727.3 |
344 |
Silent Mutation |
CAA,CAG |
Q79Q |
XP_006711790.1 |
XM_006711728.3 |
344 |
Silent Mutation |
CAA,CAG |
Q79Q |
XP_006711791.1 |
XM_011544021.2 |
344 |
Silent Mutation |
CAA,CAG |
Q79Q |
XP_011542323.1 |
XM_011544022.2 |
344 |
Silent Mutation |
CAA,CAG |
Q69Q |
XP_011542324.1 |
XM_011544023.2 |
344 |
Silent Mutation |
CAA,CAG |
Q79Q |
XP_011542325.1 |
XM_011544024.2 |
344 |
Silent Mutation |
CAA,CAG |
Q79Q |
XP_011542326.1 |
XM_011544026.2 |
344 |
Silent Mutation |
CAA,CAG |
Q79Q |
XP_011542328.1 |
XM_011544028.2 |
344 |
Silent Mutation |
CAA,CAG |
Q79Q |
XP_011542330.1 |
XM_011544029.2 |
344 |
Silent Mutation |
CAA,CAG |
Q79Q |
XP_011542331.1 |
XM_011544030.2 |
344 |
Intron |
|
|
XP_011542332.1 |
XM_017000103.1 |
344 |
Silent Mutation |
CAA,CAG |
Q79Q |
XP_016855592.1 |
XM_017000104.1 |
344 |
Silent Mutation |
CAA,CAG |
Q69Q |
XP_016855593.1 |
XM_017000105.1 |
344 |
Silent Mutation |
CAA,CAG |
Q69Q |
XP_016855594.1 |
XM_017000106.1 |
344 |
UTR 5 |
|
|
XP_016855595.1 |
XM_017000107.1 |
344 |
UTR 5 |
|
|
XP_016855596.1 |
XM_017000108.1 |
344 |
UTR 5 |
|
|
XP_016855597.1 |
XM_017000109.1 |
344 |
Silent Mutation |
CAA,CAG |
Q79Q |
XP_016855598.1 |
View Full Product Details