Product Details

SNP ID
rs201287823
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:159952330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATGGGGTTGTTGGCTCTGCAG[A/G]TGTAGGAGAGGGCACTGTCCCCCGG
Phenotype
MIM: 609738 MIM: 608589
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGSF9 PubMed Links

Gene Details

Gene
IGSF9
Gene Name
immunoglobulin superfamily member 9
There are no transcripts associated with this gene.

Gene
LINC01133
Gene Name
long intergenic non-protein coding RNA 1133
There are no transcripts associated with this gene.

Gene
SLAMF9
Gene Name
SLAM family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146172.1 713 Intron NP_001139644.1
NM_001146173.1 713 Intron NP_001139645.1
NM_033438.3 713 Missense Mutation ACC,ATC T199I NP_254273.2
XM_017002756.1 713 Missense Mutation ACC,ATC T229I XP_016858245.1

View Full Product Details