Product Details

SNP ID
rs201579975
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:999710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTACCTCTTTTCTGAGGGCGTC[C/T]AGGATGAGGGTTTTGAGCTGAGCGA
Phenotype
MIM: 608060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HES4 PubMed Links

Gene Details

Gene
HES4
Gene Name
hes family bHLH transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142467.1 463 Silent Mutation CTA,CTG L88L NP_001135939.1
NM_021170.3 463 Silent Mutation CTA,CTG L62L NP_066993.1
XM_005244771.4 463 Intron XP_005244828.1

View Full Product Details