Product Details

SNP ID
rs200986037
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:151364705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCTGCAGCATCACAGAGCCTTC[C/T]CTGGTGGAAAAGGGAATGGGGTAAG
Phenotype
MIM: 604188
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SELENBP1 PubMed Links

Gene Details

Gene
SELENBP1
Gene Name
selenium binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258288.1 1559 Silent Mutation AGA,AGG R357R NP_001245217.1
NM_001258289.1 1559 Silent Mutation AGA,AGG R461R NP_001245218.1
NM_003944.3 1559 Silent Mutation AGA,AGG R419R NP_003935.2
XM_017002757.1 1559 Intron XP_016858246.1

View Full Product Details